Product Name
Methyl 5-(4-cyanophenyl)furan-2-carboxylate 96%
Product Number
648477
Product Brand
aldrich
CAS Number
834884-75-2
Molecular Weight
227.22
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Conforms to Requirements |
Powder, Crystals and/or Chunks | |
Infrared spectrum | Conforms to Structure |
Carbon | 65.6 - 71.8 % |
Nitrogen | 5.8 - 6.4 % |
Purity (HPLC) | ≥95.5 % |