Product Name
3-Hydroxypropane-1-sulfonic acid technical, ~80% (T)
Product Number
56260
Product Brand
aldrich
CAS Number
15909-83-8
Molecular Weight
140.16
Test | Specification |
---|---|
APPEARANCE (COLOR) | Brown to Very Dark Brown |
APPEARANCE (FORM) | Liquid or Viscous Liquid |
TITRATION (T) NAOH 0.1M | ~ 80 % in water |
WATER | ~ 20 water |
CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |