Skip to Content
Merck
All Photos(1)

Documents

G005882

LC/MS/MS Analysis of Testosterone and Metabolites on Ascentis® Express C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 μm particles (53822-U)
mobile phase: [A] 5 mM ammonium formate water pH 4.0 formic acid; [B] 0.1% formic acid acetonitrile
gradient: 10 to 70% B in 5 min; held at 70% B for 3 min
flow rate: 0.6 mL/min
pressure: 3698 psi (255 bar)
column temp.: 35 °C
detector: MS, ESI(+), TIC, m/z 100-1000
injection: 5 μL
sample: 1000 ng/mL in 40:60 (water:methanol)

suitability

application for HPLC

application(s)

clinical

Analysis Note

An Ascentis Expres F5 column provided rapid, baseline resolution of testosterone and metabolites. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
    17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
    5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service