Skip to Content
Merck
All Photos(1)

Documents

76723

Supelco

1-Penten-3-one

contains ~0.1% BHT as stabilizer, analytical standard

Synonym(s):

Ethyl vinyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH=CH2
CAS Number:
Molecular Weight:
84.12
Beilstein:
1735857
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

contains

~0.1% BHT as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.419 (lit.)
n20/D 1.419

bp

38 °C/60 mmHg (lit.)

density

0.851 g/mL at 20 °C
0.845 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CCC(=O)C=C

InChI

1S/C5H8O/c1-3-5(6)4-2/h3H,1,4H2,2H3

InChI key

JLIDVCMBCGBIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Böttcher et al.
Food chemistry, 169, 492-498 (2014-09-23)
The aim of the present study was to evaluate the off-flavour masking potential of pea dextrin (PD) in emulsions rich in ω-3 and ω-6-fatty acids in comparison with maltodextrin (MD) and 2-hydroxypropyl-β-cyclodextrin (HPBCD). After optimisation of the homogenisation procedure, stable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service