Skip to Content
Merck
All Photos(1)

Key Documents

SK1M005W01

Millipore

Pellicon® XL50 Cassette with Biomax® Membrane

NMWCO 8 kDa

Synonym(s):

Pellicon® XL50 with Biomax 8 kDa Membrane, C Screen, 50 cm2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

material

polyethersulfone
polypropylene
polypropylene screen

feature

holdup volume 3.2 mL

manufacturer/tradename

Pellicon®

parameter

0.03-0.05 L/min recirculation rate
2.8 bar max. transmembrane pressure (40 psig)
5.5 bar max. inlet pressure (80 psi) at 50 °C

technique(s)

analytical sample preparation: suitable
protein purification: suitable

L

18.8 cm (7.4 in.)

W

3 cm (1.2 in.)

membrane nominal area

50 cm2

gravimetric extractables

≤7.5 mg/device

matrix

Biomax®

pore size

8 kDa NMWCO

pH

1-14(cleaning)
1-14(continuous)

General description

Device Configuration: Cassette
Filter Code: Biomax 8
Filter Type: Ultrafiltration

Features and Benefits

Type C screen (coarse screen)

Packaging

Each Pellicon® XL50 cassette is packaged one per box and includes Operating Instructions, an Accessory Kit and a Certificate of Quality

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after a water flush

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service