Skip to Content
Merck
All Photos(2)

Key Documents

70793

Millipore

BugBuster® His•Bind® Purification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

Components

•2 × 100 mlBugBuster Protein Extraction Reagent

•10,000 UBenzonase Nuclease, Purity >90%

•10 mlHis•Bind Resin

•1His•Bind Buffer Kit

•pkg/4Chromatography Columns

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS-BIND is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service