Skip to Content
Merck
All Photos(3)

Documents

05-593-I

Sigma-Aldrich

Anti-α-Dystroglycan Antibody

Synonym(s):

Dystroglycan antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

IIH6 (or IIH6C4), monoclonal

mol wt

calculated mol wt 97.03 kDa
observed mol wt ~43 and 156 kDa

species reactivity

rabbit, human, mouse

species reactivity (predicted by homology)

guinea pig, canine, rat

packaging

antibody small pack of 100 μL

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

rabbit ... DAG1(100009278)

General description

Dystroglycan (UniProt: Q28685; also known as Dystrophin-associated glycoprotein 1) is encoded by the DAG1 gene (Gene ID: 100009278) in rabbit. Dystroglycan is a component of the dystrophin glycoprotein complex that serves as an adhesion molecule and links the extracellular matrix to the intracellular actin cytoskeleton. Its expression is reported in the brain, cardiac muscle, skeletal muscle, and placenta. It is synthesized with a signal peptide (aa 1-29) that is subsequently cleaved off to generate the mature protein that is further autolytically cleaved (between 653-654) to produce a-dystroglycan (aa 30-653) and b-dystroglycan (aa 654-895). a-dystroglycan is an extracellular peripheral glycoprotein that binds to several extracellular matrix and synaptic proteins such as laminin-2 and 5, agrin, neurexin, and pikachurin. It is heavily O-glycosylated, which comprises of up to two thirds of its mass and the carbohydrate composition differs depending on tissue type. It is reported that its O-mannosyl glycosylation is required for its ligand- binding functions. b-dystroglycan is a single-pass type 1 transmembrane protein that plays important roles in connecting the extracellular matrix to the cytoskeleton and serves as a cell adhesion receptor in both muscle and non-muscle tissues. In the cell, b-dystroglycan binds to dystrophin that is linked to actin cytoskeleton. Hence, a/b dystroglycans act as a molecular axis connecting extracellular matrix with the cytoskeleton across the plasma membrane. Defects in O-mannosyl glycan have been linked to various congenital muscular dystrophies caused by aberrant a-dystroglycan glycosylation. (Ref.: Kanagawa, M., et al. (2010). J. Biol. Chem. 285(41); 31208-31216; Yamada, H., et al. (1994). FEBS Lett. 352(1); 49-53; Ervasti, JM., and Campbell, KP. (1993). J. Cell Biol. 12(4); 809-823; Ibraghimov-Beskrovnaya, O., et al. (1992). Nature. 355(6362); 696-702).

Specificity

Clone IIH6 (IIH6C4) is a mouse monoclonal antibody that detects Dystroglycan (a-Dystroglycan and uncleaved form) in multiple species.

Immunogen

Rabbit skeletal muscle membrane preparation.

Application

Quality Control Testing

Evaluated by Western Blotting in Rabbit skeletal muscle tissue lysate.

Western Blotting Analysis: 1:1,000 of this antibody detected Dystroglycan in Rabbit skeletal muscle tissue lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected Dystroglycan in Western Blotting applications (Azzag, K., et. al. (2020). Skelet Muscle. 10(1):10 ; Bijata, M., et. al. (2015). Front Cell Neurosci. 9:199).

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Dystroglycan in mouse heart and mouse skeletal muscle tissue sections.

Flow Cytometry Analysis: A representative lot detected Dystroglycan in Flow Cytometry applications (Gerin, I., et. al. (2016). Nat Commun. 7:11534).

Immunofluorescence Analysis: A representative lot detected Dystroglycan in Immunofluorescence applications (Azzag, K., et. al. (2020). Skelet Muscle. 10(1):10).

Inhibition Analysis: A representative lot reduced agrin-induced ERK activation in cardiac cells. (Bassat, E., et. al. (2017). Nature. 547(7662):179-184)

Immunohistochemistry Applications: A representative lot detected -Dystroglycan in Immunohistochemistry applications (Cataldi, M.P., et. al. (2018). Nat Commun. 9(1):3448).

Immunocytochemistry Analysis: A representative lot detected Dystroglycan in Immunocytochemistry applications (Bijata, M., et. al. (2015). Front Cell Neurosci. 9:199).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-a-Dystroglycan, clone IIH6C4, Cat. No. 05-593-I, is a mouse monoclonal antibody that detects Dystroglycan and is tested for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence Inhibition, Immunohistochemistry, and Western Blotting.

Physical form

Mouse monoclonal antibody without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service