Skip to Content
Merck
All Photos(1)

Key Documents

ENAH95E741A2

Enamine

4-methoxy-2-methyl-3-nitrobenzene-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
265.67
MDL number:

Iupac Name

4-methoxy-2-methyl-3-nitrobenzenesulfonyl chloride

Inchi Code

1S/C8H8ClNO5S/c1-5-7(16(9,13)14)4-3-6(15-2)8(5)10(11)12/h3-4H,1-2H3

InChI key

GJUNPTLHMBQUNJ-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-27158349

MSDS

https://www.enaminestore.com/catalog/EN300-27158349


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service