Skip to Content
Merck
All Photos(1)

Key Documents

ATC311768311

Atlantic Research Chemicals Ltd

tert-Butyl 4-[(5-Nitropyridin-2-yl)oxy]piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
323.35
MDL number:

Iupac Name

tert-butyl 4-[(5-nitro-2-pyridinyl)oxy]-1-piperidinecarboxylate

Inchi Code

1S/C15H21N3O5/c1-15(2,3)23-14(19)17-8-6-12(7-9-17)22-13-5-4-11(10-16-13)18(20)21/h4-5,10,12H,6-9H2,1-3H3

InChI key

JVQMZHDBFJVUEO-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=XS002286

MSDS

https://atlantic-chemicals.com/product/?code=XS002286


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service