Skip to Content
Merck
All Photos(1)

Key Documents

ATE014416657

AstaTech, Inc.

4-N-PHTHALOYLGLYAMINOMETHYL-CYCLOHEXANONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
257.29
MDL number:

Iupac Name

2-[(4-oxocyclohexyl)methyl]-1H-isoindole-1,3(2H)-dione

Inchi Code

1S/C15H15NO3/c17-11-7-5-10(6-8-11)9-16-14(18)12-3-1-2-4-13(12)15(16)19/h1-4,10H,5-9H2

InChI key

CIXKYGZRROIQLQ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

96%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=5815

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=59325


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service