Skip to Content
Merck
All Photos(2)

Documents

Z741826

Extech Non-Contact Forehead InfraRed Thermometer

Sign Into View Organizational & Contract Pricing


About This Item

description

Optimum measurement distance of 1.9" to 5.9" (5 to 15cm)

feature

CE compliant
accuracy± 0.5 °F (±0.3°C)
battery included (2 AA)
resolution 0.1 °F/C
(WEEE Compliant)

packaging

pkg of 1 ea

manufacturer/tradename

Extech Instruments IR200

temperature measuring range

89.6-108.5 °F ((32.0°C to 42.5°C)

size

6.3 in. × 3.2 in. × 1.7 in., 160x82x42 mm

weight

6.24 oz177 g

Features and Benefits

  • Adjustable alarm alerts user visually and audibly when temperature exceeds programmed limit
  • Measures surface temperature from 32°F to 140°F (0.0°C to 60.0°C)
  • Fast response (0.5 seconds)
  • Simply press the trigger and read temperature on the large backlit LCD display
  • Memory stores up to 32 readings for easy recall
  • Complete with 2 AA batteries and pouch


Applications:
  • Measure body (forehead) temperature
  • Monitor individuals for temperature change
  • Non-contact technique reduces potential

spread of diseases from using contact devices

Note: This thermometer is intended for scanning individuals or monitoring an individual for potential elevated temperatures. It is not a substitute for a clinical thermometer. Always use a clinical thermometer when high accuracy body temperature measurements are required. IR Thermometers that detect elevated body temperature will vary with various factors and should not be relied upon as the sole determinant of a person′s body temperature. Use of additional medical devices and/or healthcare professionals will be needed to properly diagnose the condition of persons in any health screening assessment to identify elevated body temperature for any person.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service