GC Analysis of Bleed Profiles of GC Septa on SLB®-5ms
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
40 °C (3 min), 15 °C/min to 325 °C (15 min)
inj. temp.
250 °C
detector
FID, 325 °C
carrier gas
helium, 25 cm/sec constant
injection
1 μL splitless (1 min)
liner
4 mm I.D., single taper, unpacked
sample
solvent extracts of septa, dodecane (as an internal standard) added to 100 μg/mL
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany