Merck
  • Cytocompatible polyion complex gel of poly(Pro-Hyp-Gly) for simultaneous rat bone marrow stromal cell encapsulation.
Cytocompatible polyion complex gel of poly(Pro-Hyp-Gly) for simultaneous rat bone marrow stromal cell encapsulation.
Journal of biomaterials science. Polymer edition (2017-05-19)
Farah Nurlidar, Mime Kobayashi, Kayo Terada, Tsuyoshi Ando, Masao Tanihara
ABSTRACT
Polyion complex (PIC) gel of poly(Pro-Hyp-Gly) was successfully fabricated by simply mixing polyanion and polycation derivatives of poly(Pro-Hyp-Gly), a collagen-like polypeptide. The polyanion, succinylated poly(Pro-Hyp-Gly), and the polycation, arginylated poly(Pro-Hyp-Gly), contain carboxy (pK
Sigma-Aldrich
L-Arginine methyl ester dihydrochloride, ≥98.0% (AT)
Pricing and availability is not currently available.