Merck
  • In vitro and in vivo studies of cholinesterases and carboxylesterases in Planorbarius corneus exposed to a phosphorodithioate insecticide: Finding the most sensitive combination of enzymes, substrates, tissues and recovery capacity.
In vitro and in vivo studies of cholinesterases and carboxylesterases in Planorbarius corneus exposed to a phosphorodithioate insecticide: Finding the most sensitive combination of enzymes, substrates, tissues and recovery capacity.
Aquatic toxicology (Amsterdam, Netherlands) (2016-10-11)
Sofía Otero, Gisela Kristoff
ABSTRACT
Organophosphate insecticides (OPs) continue to be an important class of agrochemicals used in modern agriculture worldwide. Even though these pesticides persist in the environment for a relatively short time, they show a high acute toxicity that may represent a serious hazard for wildlife. Sub-lethal effects on non-target species are a focus in pest management programs and should be used as biomarkers. Cholinesterases (ChEs) are the most used biomarker of OP exposure in vertebrate and invertebrate species. However, the combined monitoring of ChE and carboxylesterase (CE) activities may provide a more useful indication of exposure and effect of the organisms. The objective of the present work was to find the most sensitive combination of enzyme, substrate, tissue and capacity to recovery of B-esterases in the freshwater gastropod Planorbarius corneus exposed to the OP azinphos-methyl. For this purpose, ChE and CE activities in different tissues of P. corneus (head-foot, pulmonary region, digestive gland, gonads and whole organism soft tissue) were studied. Measurements of ChE activity were performed using three substrates: acetylthiocholine, propionylthiocholine and butyrylthiocholine and CE activity using four different substrates: p-nitrophenyl acetate, p-nitrophenyl butyrate, 1-naphthyl acetate, and 2-naphthyl acetate in control and exposed organisms. Finally, the recovery rates of ChE and CE activities following 48h exposure to azinphos-methyl were analyzed. Our results show a preference for acetylthiocholine as substrate, a high inhibition with eserine (a selective ChE inhibitor) and inhibition with excess of substrate in all the analyzed tissues. The highest ChE and CE activity was found in the pulmonary region and in the digestive gland, respectively. The highest CE V
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
2-Naphthyl acetate
Sigma-Aldrich
Propionylthiocholine iodide, ≥98%