Merck
  • Defective DNA single-strand break repair is responsible for senescence and neoplastic escape of epithelial cells.
Defective DNA single-strand break repair is responsible for senescence and neoplastic escape of epithelial cells.
Nature communications (2016-01-30)
Joe Nassour, Sébastien Martien, Nathalie Martin, Emeric Deruy, Elisa Tomellini, Nicolas Malaquin, Fatima Bouali, Laure Sabatier, Nicolas Wernert, Sébastien Pinte, Eric Gilson, Albin Pourtier, Olivier Pluquet, Corinne Abbadie
ABSTRACT
The main characteristic of senescence is its stability which relies on the persistence of DNA damage. We show that unlike fibroblasts, senescent epithelial cells do not activate an ATM-or ATR-dependent DNA damage response (DDR), but accumulate oxidative-stress-induced DNA single-strand breaks (SSBs). These breaks remain unrepaired because of a decrease in PARP1 expression and activity. This leads to the formation of abnormally large and persistent XRCC1 foci that engage a signalling cascade involving the p38MAPK and leading to p16 upregulation and cell cycle arrest. Importantly, the default in SSB repair also leads to the emergence of post-senescent transformed and mutated precancerous cells. In human-aged skin, XRCC1 foci accumulate in the epidermal cells in correlation with a decline of PARP1, whereas DDR foci accumulate mainly in dermal fibroblasts. These findings point SSBs as a DNA damage encountered by epithelial cells with aging which could fuel the very first steps of carcinogenesis.
Sigma-Aldrich
3-Aminobenzamide, ≥99% (TLC)
Sigma-Aldrich
Catalase−polyethylene glycol, lyophilized powder, ~40,000 units/mg protein
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder