Merck
  • A soluble form of nano-sized colloidal manganese(IV) oxide as an efficient catalyst for water oxidation.
A soluble form of nano-sized colloidal manganese(IV) oxide as an efficient catalyst for water oxidation.
Dalton transactions (Cambridge, England : 2003) (2011-03-02)
Mohammad Mahdi Najafpour
ABSTRACT
A soluble form of colloidal manganese(IV) oxide showed efficient oxygen evolution or water oxidation in presence of oxone, H(2)O(2), cerium(IV) ammonium nitrate and tris(2,2'-bipyridyl)ruthenium(III).
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, ReagentPlus®, ≥99%