Merck
The analysis of total and form-variable tin in various media.
Neurotoxicology (1984-01-01)
P Mushak
ABSTRACT
Methods for both total and form-variable tin analysis in biological media are reviewed. While total tin analysis was common in the past, and in some cases still is, better understanding of the toxicology and toxicokinetics of organotins in biological systems requires knowledge of specific tin forms present. Both chromatographic and non-chromatographic methods for chemical speciation of organotins in various media have been described, the former appearing to offer the most promising approaches. High performance liquid chromatography (HPLC) in tandem with tin-specific metal detection presently appears to offer the most promise.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Tin, foil, 4mm disks, thickness 0.025mm, as rolled, 98.8%
Tin, foil, 25mm disks, thickness 0.075mm, as rolled, 98.8%
Tin, foil, 15mm disks, thickness 0.075mm, as rolled, 98.8%
Tin, foil, 4mm disks, thickness 0.025mm, as rolled, 99.95%
Tin, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Tin, foil, 5m coil, thickness 0.0125mm, 97.4%
Tin, foil, 10m coil, thickness 0.007mm, 97.4%
Tin, foil, 2m coil, thickness 0.01mm, 97.4%
Tin, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Tin, foil, 4mm disks, thickness 0.008mm, 99.75%
Tin, foil, 25mm disks, thickness 0.05mm, as rolled, 98.8%
Tin, foil, 300x300mm, thickness 0.20mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.008mm, 99.75%
Tin, foil, 25mm disks, thickness 0.25mm, as rolled, 98.8%
Tin, foil, 50mm disks, thickness 0.025mm, as rolled, 98.8%
Tin, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 1m coil, thickness 0.072mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 2m coil, thickness 0.125mm, as rolled, 98.8%
Tin, foil, 4mm disks, thickness 0.009mm, 97.4%
Tin, foil, 10mm disks, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 0.5m coil, thickness 0.01mm, 97.4%