Merck
  • Novel N-benzoyl-2-hydroxybenzamide disrupts unique parasite secretory pathway.
Novel N-benzoyl-2-hydroxybenzamide disrupts unique parasite secretory pathway.
Antimicrobial agents and chemotherapy (2012-02-23)
Alina Fomovska, Qingqing Huang, Kamal El Bissati, Ernest J Mui, William H Witola, Gang Cheng, Ying Zhou, Caroline Sommerville, Craig W Roberts, Sam Bettis, Sean T Prigge, Gustavo A Afanador, Mark R Hickman, Patty J Lee, Susan E Leed, Jennifer M Auschwitz, Marco Pieroni, Jozef Stec, Stephen P Muench, David W Rice, Alan P Kozikowski, Rima McLeod
ABSTRACT
Toxoplasma gondii is a protozoan parasite that can damage the human brain and eyes. There are no curative medicines. Herein, we describe our discovery of N-benzoyl-2-hydroxybenzamides as a class of compounds effective in the low nanomolar range against T. gondii in vitro and in vivo. Our lead compound, QQ-437, displays robust activity against the parasite and could be useful as a new scaffold for development of novel and improved inhibitors of T. gondii. Our genome-wide investigations reveal a specific mechanism of resistance to N-benzoyl-2-hydroxybenzamides mediated by adaptin-3β, a large protein from the secretory protein complex. N-Benzoyl-2-hydroxybenzamide-resistant clones have alterations of their secretory pathway, which traffics proteins to micronemes, rhoptries, dense granules, and acidocalcisomes/plant-like vacuole (PLVs). N-Benzoyl-2-hydroxybenzamide treatment also alters micronemes, rhoptries, the contents of dense granules, and, most markedly, acidocalcisomes/PLVs. Furthermore, QQ-437 is active against chloroquine-resistant Plasmodium falciparum. Our studies reveal a novel class of compounds that disrupts a unique secretory pathway of T. gondii, with the potential to be used as scaffolds in the search for improved compounds to treat the devastating diseases caused by apicomplexan parasites.
Sigma-Aldrich
4-Ethylbenzoyl chloride, 97%