Merck
  • Chronic systemic D-galactose exposure induces memory loss, neurodegeneration, and oxidative damage in mice: protective effects of R-alpha-lipoic acid.
Chronic systemic D-galactose exposure induces memory loss, neurodegeneration, and oxidative damage in mice: protective effects of R-alpha-lipoic acid.
Journal of neuroscience research (2006-05-20)
Xu Cui, Pingping Zuo, Qing Zhang, Xuekun Li, Yazhuo Hu, Jiangang Long, Lester Packer, Jiankang Liu
ABSTRACT
Chronic systemic exposure of mice, rats, and Drosophila to D-galactose causes the acceleration of senescence and has been used as an aging model. The underlying mechanism is yet unclear. To investigate the mechanisms of neurodegeneration in this model, we studied cognitive function, hippocampal neuronal apoptosis and neurogenesis, and peripheral oxidative stress biomarkers, and also the protective effects of the antioxidant R-alpha-lipoic acid. Chronic systemic exposure of D-galactose (100 mg/kg, s.c., 7 weeks) to mice induced a spatial memory deficit, an increase in cell karyopyknosis, apoptosis and caspase-3 protein levels in hippocampal neurons, a decrease in the number of new neurons in the subgranular zone in the dentate gyrus, a reduction of migration of neural progenitor cells, and an increase in death of newly formed neurons in granular cell layer. The D-galactose exposure also induced an increase in peripheral oxidative stress, including an increase in malondialdehyde, a decrease in total anti-oxidative capabilities (T-AOC), total superoxide dismutase (T-SOD), and glutathione peroxidase (GSH-Px) activities. A concomitant treatment with lipoic acid ameliorated cognitive dysfunction and neurodegeneration in the hippocampus, and also reduced peripheral oxidative damage by decreasing malondialdehyde and increasing T-AOC and T-SOD, without an effect on GSH-Px. These findings suggest that chronic D-galactose exposure induces neurodegeneration by enhancing caspase-mediated apoptosis and inhibiting neurogenesis and neuron migration, as well as increasing oxidative damage. In addition, D-galactose-induced toxicity in mice is a useful model for studying the mechanisms of neurodegeneration and neuroprotective drugs and agents.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit