Merck
  • Application of (13)C-stable isotope probing to identify RDX-degrading microorganisms in groundwater.
Application of (13)C-stable isotope probing to identify RDX-degrading microorganisms in groundwater.
Environmental pollution (Barking, Essex : 1987) (2013-04-23)
Kun-Ching Cho, Do Gyun Lee, Hyungkeun Roh, Mark E Fuller, Paul B Hatzinger, Kung-Hui Chu
ABSTRACT
We employed stable isotope probing (SIP) with (13)C-labeled hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) to identify active microorganisms responsible for RDX biodegradation in groundwater microcosms. Sixteen different 16S rRNA gene sequences were derived from microcosms receiving (13)C-labeled RDX, suggesting the presence of microorganisms able to incorporate carbon from RDX or its breakdown products. The clones, residing in Bacteroidia, Clostridia, α-, β- and δ-Proteobacteria, and Spirochaetes, were different from previously described RDX degraders. A parallel set of microcosms was amended with cheese whey and RDX to evaluate the influence of this co-substrate on the RDX-degrading microbial community. Cheese whey stimulated RDX biotransformation, altered the types of RDX-degrading bacteria, and decreased microbial community diversity. Results of this study suggest that RDX-degrading microorganisms in groundwater are more phylogenetically diverse than what has been inferred from studies with RDX-degrading isolates.
Product Description
Supelco
RDX solution, 1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®