Merck
Patch testing with textile allergens: the mayo clinic experience.
Dermatitis : contact, atopic, occupational, drug (2012-11-22)
Ashley B Wentworth, Donna M Richardson, Mark D P Davis
ABSTRACT
Recognition of allergic contact dermatitis attributed to textile dyes and resins is steadily increasing. This study aims to review the results of patch testing with a textile series at our institution and to compare with previously published reports. We performed a retrospective review of results in patients who underwent patch testing using a series of textile dyes and resins from January 1, 2000, through September 30, 2011. A total of 671 patients (mean age, 56.5 years; female, 65.9%) were patch tested with the textile series (42 dyes and resins). These patients were also generally tested with the standard patch test series (n = 620). Of the patients, 219 (32.6%) demonstrated allergic reaction to 1 or more textile dyes and resins, and 71 (10.6%) manifested irritant reactions. The most frequent allergens were disperse blue 106 1% (8.3%), disperse blue 124 1% (8.0%), and melamine formaldehyde 7% (8.0%). Of patients tested with the standard series, 36 (5.8%) showed a positive reaction to the traditional textile screening allergen p-phenylenediamine 1%. Supplementing the standard series with the textile series increased detection of textile allergies. In patients suspected of textile allergy, addition of the textile series is necessary for appropriate diagnosis.
Product Description
Supelco
Disperse Blue 124