Merck
A clay-vesicle system for water purification from organic pollutants.
Water research (2007-10-05)
Tomas Undabeytia, Shlomo Nir, Trinidad Sánchez-Verdejo, Jaime Villaverde, Celia Maqueda, Esmeralda Morillo
ABSTRACT
Vesicle-clay complexes in which positively charged vesicles composed of didodecyldimethylammonium bromide (DDAB) were adsorbed on montmorillonite removed efficiently anionic (sulfentrazone, imazaquin) and neutral (alachlor, atrazine) pollutants from water. These complexes (0.5% w:w) removed 92-100% of sulfentrazone, imazaquin and alachlor and 60% of atrazine from a solution containing 10mg/L of it. A synergistic effect on the adsorption of atrazine was observed when all pollutants were present simultaneously (30 mg/L each), its percentage of removal being 85.5. Column filters (18 cm) filled with a mixture of quartz sand and vesicle-clay (100:1, w:w) were tested. For the passage of 1L (25 pore volumes) of a solution including all the pollutants at 10mg/L each, removal was complete for sulfentrazone and imazaquin, 94% for alachlor and 53.1% for atrazine, whereas removal was significantly less efficient when using activated carbon. A similar advantage of the vesicle-clay filter was observed for the capacities of removal.
Product Description
Supelco
Imazaquin, PESTANAL®, analytical standard