Merck
  • Interleukin-24 attenuates β-glycerophosphate-induced calcification of vascular smooth muscle cells by inhibiting apoptosis, the expression of calcification and osteoblastic markers, and the Wnt/β-catenin pathway.
Interleukin-24 attenuates β-glycerophosphate-induced calcification of vascular smooth muscle cells by inhibiting apoptosis, the expression of calcification and osteoblastic markers, and the Wnt/β-catenin pathway.
Biochemical and biophysical research communications (2012-10-16)
Ki-Mo Lee, Haeng-A Kang, Min Park, Hwa-Youn Lee, Ha-Rim Choi, Chul-Ho Yun, Jae-Wook Oh, Hyung-Sik Kang
ABSTRACT
Vascular calcification is a hallmark of cardiovascular disease. Interleukin-24 (IL-24) has been known to suppress tumor progression in a variety of human cancers. However, the role of IL-24 in the pathophysiology of diseases other than cancer is unclear. We investigated the role of IL-24 in vascular calcification. IL-24 was applied to a β-glycerophosphate (β-GP)-induced rat vascular smooth muscle cell (VSMC) calcification model. In this study, IL-24 significantly inhibited β-GP-induced VSMC calcification, as determined by von Kossa staining and calcium content. The inhibitory effect of IL-24 on VSMC calcification was due to the suppression of β-GP-induced apoptosis and expression of calcification and osteoblastic markers. In addition, IL-24 abrogated β-GP-induced activation of the Wnt/β-catenin pathway, which plays a key role in the pathogenesis of vascular calcification. The specificity of IL-24 for the inhibition of VSMC calcification was confirmed by using a neutralizing antibody to IL-24. Our results suggest that IL-24 inhibits β-GP-induced VSMC calcification by inhibiting apoptosis, the expression of calcification and osteoblastic markers, and the Wnt/ β-catenin pathway. Our study may provide a novel mechanism of action of IL-24 in cardiovascular disease and indicates that IL-24 is a potential therapeutic agent in VSMC calcification.
Sigma-Aldrich
Glycerol phosphate calcium salt, >98% (TLC)