Merck
  • Role of endogenous hydrogen sulfide on renal damage induced by adriamycin injection.
Role of endogenous hydrogen sulfide on renal damage induced by adriamycin injection.
Archives of toxicology (2011-05-19)
Heloísa Della Coletta Francescato, Evelyn Cristina Santana Marin, Fernando de Queiroz Cunha, Roberto Silva Costa, Cleonice Giovanini Alves da Silva, Terezila Machado Coimbra
ABSTRACT
A single injection of adriamycin (ADR) induces marked and persistent proteinuria in rats that progress to glomerular and tubulointerstitial lesions. It has been shown that ADR-induced nephrotoxicity is mediated, at least in part, by oxidative stress that lead to inflammation. Endogenous hydrogen sulfide (H₂S) is synthesized from L-cysteine and is an important signaling molecule in inflammation. This study evaluates the effect of DL-propargylglycine (PAG), an inhibitor of endogenous H₂S formation, on the evolution of renal damage induced by ADR. The rats were injected i.p. with 0.15 M NaCl or PAG (50 mg/kg) 2 h after ADR injection (3.5 mg/kg). Control rats were injected with 0.15 M NaCl or PAG only. Twenty hours urine samples were collected for albuminuria and creatinine measurements on days 1 and 14 after saline or ADR injections and on days 2 and 15 blood samples were collected to measure plasma creatinine, then the rats were killed. The kidneys were removed for H₂S formation evaluation, renal lipid peroxidation and glutathione levels, and histological and immunohistochemical analysis. On day 2 after ADR injection the rats presented increase in oxidative stress associated with neutrophils and macrophages influx in renal tissue. On day 15 the rats also presented increased desmin expression at glomerular edge and vimentin in cortical tubulointerstitium, as well as albuminuria. All these alterations were reduced by PAG injection. The protective effect of PAG on ADR nephrotoxicity was associated to decreased H₂S formation and to restriction of oxidative stress and inflammation in the renal cortex.
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor
Sigma-Aldrich
L-C-Propargylglycine, ≥99.0% (TLC)