Merck
  • Preparation of Fe oxide nanoparticles for environmental applications: arsenic removal.
Preparation of Fe oxide nanoparticles for environmental applications: arsenic removal.
Environmental geochemistry and health (2010-04-14)
Ulker Beker, Luis Cumbal, Dilek Duranoglu, Ilknur Kucuk, Arup K Sengupta
ABSTRACT
The objective of this study is to examine the adsorption-desorption behavior of a magnetically active hybrid sorbent (MAHS) material, prepared by dispersing colloid-like hydrated iron oxide particles in the outer periphery of a macroporous ion-exchange resin (Amberlite XAD-2). The experimental results show that the new sorbent material can simultaneously remove arsenic (V) and a chlorinated organic compound (2,6-dichlorophenol [2,6-DCP]) from aqueous solutions at around neutral pH. The recovery of arsenic and 2,6-DCP from MAHS was conducted using a regenerant containing 50% (v/v) CH3OH + 3% (w/v) NaOH. In less than 10 bed volumes of regenerant, more than 90% of As(V) and 2,6-DCP were recovered.
Sigma-Aldrich
2,6-Dichlorophenol, 99%