Merck
  • [Microdetermination of proteins with Titan yellow by the resonance light scattering method].
[Microdetermination of proteins with Titan yellow by the resonance light scattering method].
Guang pu xue yu guang pu fen xi = Guang pu (2005-10-06)
Su-ling Feng, Jin Wang, Jing Fan
ABSTRACT
A resonance light scattering method for the determination of trace proteins was developed. In the presence of Triton X-100, proteins reacted with Titan yellow to form a combination product, resulting a significant enhancement of resonance light scattering (RLS). The deltaI(RLS) was directly proportional to the concentration of protein in the range of 0.03-0.9 microg x mL(-1), with the detection limit 17.7 ng x mL(-1) for BSA. This method was applied to the determination of the proteins in synthetic and human serum samples, and compared to the CBB method, with satisfactory results.
Sigma-Aldrich
Thiazole Yellow G, adsorption indicator, for microscopy (fluorescence indicator)