Merck
  • Heterogeneity of tyrosine hydroxylase expressing neurons in the main olfactory bulb of the mouse.
Heterogeneity of tyrosine hydroxylase expressing neurons in the main olfactory bulb of the mouse.
Neuroscience research (2019-10-21)
Toshio Kosaka, Angela Pignatelli, Katsuko Kosaka
ABSTRACT
The structural features of dopamine (DA)-GABAergic neurons in the mouse main olfactory bulbs were examined, using both wild type and transgenic TH-GFP mice, with the combination of several methods; the immunocytochemistry, biotinylated dextran amine labeling, lucifer yellow injection in fixed slices, biocytin injection in live slice and the functional olfactory deprivation. DA-GABAergic neurons were clustered in the glomerular layer (GL) but they also scattered in other layers. DA-GABAergic juxtaglomerular neurons, were classified into 5 groups based on their structural features and named as follows: 1) Large periglomerular (LPG) cells with tuft-like glomerular dendritic branches and apparent axons extending to the distant glomeruli, which correspond to the " inhibitory juxtaglomerular association (IJGA) neurons" participating in the interglomerular association system. 2) Small periglomerular (SPG) cells including both axonic and anaxonic ones; the axonic SPG cells might correspond to the classical periglomerular cells. 3) Transglomerular cells extending dendritic processes spanning 2 or more glomeruli. 4) Incrusting cells extending their dendritic branches mainly in the periphery of the glomeruli. 5) Other various neurons not-yet classified. In the layers other than the GL various types of TH expressing neurons were scattered; some of them extended dendritic processes into the GL.
Sigma-Aldrich
Anti-Glutamic Acid Decarboxylase 65 (5-22) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from sheep