Merck
  • Purification and properties of a protamine kinase from bovine kidney microsomes.
Purification and properties of a protamine kinase from bovine kidney microsomes.
Archives of biochemistry and biophysics (1992-08-15)
G D Amick, S A Reddy, Z Damuni
ABSTRACT
About an eightfold increase in protamine kinase activity was detected following extraction of highly purified microsomes from bovine kidney with 1% Triton X-100. Relative to the soluble fraction, the microsomes contained about 30% protamine kinase activity. The microsomal protamine kinase was purified to apparent homogeneity. The purified enzyme exhibited an apparent M(r) approximately 45,000 as estimated by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and by gel permeation chromatography on Sephacryl S-200. Relative to protamine, the purified kinase exhibited about 100% activity with the synthetic peptide RRLSSLRA and about 5, 8, and less than 0.1% activity with casein, histone H2B, and histone H1, respectively. The purified kinase phosphorylated several 40 S ribosome polypeptides. One of these polypeptides was identified as ribosomal protein S6 by N-terminal sequencing. About 2.5 mol of phosphoryl groups was incorporated per mole of ribosomal protein S6 following incubation of the 40 S ribosomes with the purified kinase. Following incubation with protein phosphatase 2A2, purified preparations of the protamine kinase were inactivated. These properties were identical to those of purified preparations of a protamine kinase from extracts of bovine kidney cytosol (Z. Damuni, G.D. Amick, and T.R. Sneed, 1989, J. Biol. Chem. 264, 6412-6418). Near identical peptide patterns were obtained following incubation of purified preparations of the microsomal and cytosolic protamine kinases with Staphylococcus aureus V8 proteinase. The results indicate that a form of the cytosolic protamine kinase is present in microsomes.
Sigma-Aldrich
Protein Phosphatase 2A2 from bovine kidney, buffered aqueous glycerol solution