Merck
  • Gefitinib-loaded DSPE-PEG2000 nanomicelles with CD133 aptamers target lung cancer stem cells.
Gefitinib-loaded DSPE-PEG2000 nanomicelles with CD133 aptamers target lung cancer stem cells.
World journal of surgical oncology (2017-09-01)
Xiaolong Huang, Jingsong Huang, Dewen Leng, Shuo Yang, Qi Yao, Jin Sun, Jun Hu
ABSTRACT
Lung cancer stem cells (CSCs) are considered to be the seed of lung cancer, and CD133 is a marker of lung CSCs. Here, we developed gefitinib-loaded poly(ethylene glycol) 2000-distearoylphosphatidylethanolamine nanomicelles with CD133 aptamers (M-Gef-CD133) to eliminate CD133+ lung CSCs. M-Gef-CD133 was prepared using a lipid-film-based approach. The targeting and activity of M-Gef-CD133 towards lung CSCs were evaluated. M-Gef-CD133 were small (25 nm) and showed enhanced cytotoxic effect towards CD133+ lung CSCs compared with non-targeted M-Gef and gefitinib. Notably, M-Gef-CD133 could significantly reduce tumor sphere formation and the percentage of CD133+ lung CSCs, indicating that it possesses selective toxicity against CD133+ lung CSCs. The interaction of CD133 aptamers and CD133 shows promise in the delivery of gefitinib to CD133+ lung CSCs, and M-Gef-CD133 represents a promising treatment to target lung CSCs.