Merck
  • Identification of evolutionarily conserved gene networks mediating neurodegenerative dementia.
Identification of evolutionarily conserved gene networks mediating neurodegenerative dementia.
Nature medicine (2018-12-05)
Vivek Swarup, Flora I Hinz, Jessica E Rexach, Ken-Ichi Noguchi, Hiroyoshi Toyoshiba, Akira Oda, Keisuke Hirai, Arjun Sarkar, Nicholas T Seyfried, Chialin Cheng, Stephen J Haggarty, Murray Grossman, Vivianna M Van Deerlin, John Q Trojanowski, James J Lah, Allan I Levey, Shinichi Kondou, Daniel H Geschwind
ABSTRACT
Identifying the mechanisms through which genetic risk causes dementia is an imperative for new therapeutic development. Here, we apply a multistage, systems biology approach to elucidate the disease mechanisms in frontotemporal dementia. We identify two gene coexpression modules that are preserved in mice harboring mutations in MAPT, GRN and other dementia mutations on diverse genetic backgrounds. We bridge the species divide via integration with proteomic and transcriptomic data from the human brain to identify evolutionarily conserved, disease-relevant networks. We find that overexpression of miR-203, a hub of a putative regulatory microRNA (miRNA) module, recapitulates mRNA coexpression patterns associated with disease state and induces neuronal cell death, establishing this miRNA as a regulator of neurodegeneration. Using a database of drug-mediated gene expression changes, we identify small molecules that can normalize the disease-associated modules and validate this experimentally. Our results highlight the utility of an integrative, cross-species network approach to drug discovery.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Anti-VSNL1 Antibody, clone 2D11, clone 2D11, from mouse
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture