
Zinc trifluoromethanesulfinate

Synonym(s):
1,1,1-Trifluoro-methanesulfinic acid zinc salt (2:1), Baran trifluoromethylation reagent, Bis(((trifluoromethyl)sulfinyl)oxy)zinc, TFMS
Empirical Formula (Hill Notation):
C2F6O4S2Zn
CAS Number:
Molecular Weight:
331.53
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Reaction Type
Reagent Type
Showing 1-1 of 1