
Zidovudine impurity standard

Synonym(s):
3′-Azido-3′-deoxythymidine, AZT, Azidothymidine, ZDV, Zidovudine
Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing