Skip to Content
Merck
trans-(±)-ACPD monohydrate solid, ≥97% (NMR)

trans-(±)-ACPD monohydrate

trans-(±)-ACPD monohydrate solid, ≥97% (NMR)
Synonym(s):
trans-(±)-1-Amino-1,3-cyclopentanedicarboxylic acid monohydrate
Empirical Formula (Hill Notation):
C7H11NO4 · H2O
CAS Number:
Molecular Weight:
191.18

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing