![Thiamine nitrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/382/5cb342ab-00ad-4786-9686-a465229b7a6b/640/5cb342ab-00ad-4786-9686-a465229b7a6b.png)
Thiamine Nitrate
![Thiamine nitrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/382/5cb342ab-00ad-4786-9686-a465229b7a6b/640/5cb342ab-00ad-4786-9686-a465229b7a6b.png)
Synonym(s):
Thiamine nitrate, Vitamin B1 nitrate
Empirical Formula (Hill Notation):
C12H17N5O4S
CAS Number:
Molecular Weight:
327.36
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2