
Sodium calcium edetate

Synonym(s):
Ethylenediaminetetraacetic acid calcium disodium salt, (Ethylenedinitrilo)tetraacetic acid calcium disodium salt, EDTA calcium disodium salt, Edathamil
Linear Formula:
C10H12N2O8CaNa2
CAS Number:
Molecular Weight:
374.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing