![Quercetin United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/802/b03c7abe-0972-452e-a6a2-dffee95f8e54/640/b03c7abe-0972-452e-a6a2-dffee95f8e54.png)
Quercetin
![Quercetin United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/802/b03c7abe-0972-452e-a6a2-dffee95f8e54/640/b03c7abe-0972-452e-a6a2-dffee95f8e54.png)
Synonym(s):
Quercetin dihydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one dihydrate, 3,3′,4′,5,7-Pentahydroxyflavone dihydrate
Empirical Formula (Hill Notation):
C15H10O7 · 2H2O
CAS Number:
Molecular Weight:
338.27
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing