PYREXPLUS® narrow-mouth reagent bottle, with Pyrex® ST stopper, protective coating
Synonym(s):
Corning® 61500
Products
Product Category
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Showing 1-5 of 5