Skip to Content
Merck
vibrant-m

Oxaliplatin Related Compound B

vibrant-m
Synonym(s):
[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N′)dinitratoplatinum(II)
Empirical Formula (Hill Notation):
C6H14N4O6Pt
CAS Number:
Molecular Weight:
433.28

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1