
Luperox® 331M80, 1,1-Bis(tert-butylperoxy)cyclohexane solution

Synonym(s):
1,1-Bis(tert-butylperoxy)cyclohexane solution
Linear Formula:
C6H10[OOC(CH3)3]2
CAS Number:
Molecular Weight:
260.37
Products
Brand
Formula Weight
Melting Point (°C)
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing