
Lactitol

Synonym(s):
D-Lactitol monohydrate, 4-O-β-D-Galactopyranosyl-D-glucitol
Empirical Formula (Hill Notation):
C12H24O11 · H2O
CAS Number:
Molecular Weight:
362.33
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing