
JumpStart™ REDTaq® ReadyMix™ Reaction Mix

Products
Product Category
Brand
Color
Physical Form
Number of Reactions
Quality Segment
dNTPs
Hot Start
PCR Application
Application
Sample Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing