
Indomethacin Related Compound A

Synonym(s):
5-Methoxy-2-methyl-3-indoleacetic acid, 2-(5-Methoxy-2-methyl-1H-indol-3-yl)acetic acid
Empirical Formula (Hill Notation):
C12H13NO3
CAS Number:
Molecular Weight:
219.24
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing