
Ethyl tridecanoate

Synonym(s):
Tridecanoic acid ethyl ester, Ethyl tridecanoate
Linear Formula:
CH3(CH2)11COOC2H5
CAS Number:
Molecular Weight:
242.40
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing