
Endfitting w/ Fixed 10 μm Pyrex® Frit for 20 mm ID Glass TSKgel® Columns

Synonym(s):
TSKgel® Hardware and Accessories
Products
Manufacturer
Product Line
Package Size
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing