
Eletriptan hydrobromide

Empirical Formula (Hill Notation):
C22H26N2O2S · HBr
CAS Number:
Molecular Weight:
463.43
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing