Skip to Content
Merck
EHop-016 ≥98% (HPLC)

EHop-016

EHop-016 ≥98% (HPLC)
Synonym(s):
N4-(9-Ethyl-9H-carbazol-3-yl)-N2-[3-(4-morpholinyl)propyl]-2,4-pyrimidinediamine
Empirical Formula (Hill Notation):
C25H30N6O
CAS Number:
Molecular Weight:
430.55

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing