![D-Valacyclovir certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/306/933/89cfc91d-2445-4ad2-a8b2-cf2f4e5f546b/640/89cfc91d-2445-4ad2-a8b2-cf2f4e5f546b.jpg)
D-Valacyclovir
![D-Valacyclovir certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/306/933/89cfc91d-2445-4ad2-a8b2-cf2f4e5f546b/640/89cfc91d-2445-4ad2-a8b2-cf2f4e5f546b.jpg)
Synonym(s):
D-Valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester
Empirical Formula (Hill Notation):
C13H20N6O4
CAS Number:
Molecular Weight:
324.34
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2