
Cyclopentyl(phenyl)methanone

Synonym(s):
Cyclopentyl phenyl ketone
Empirical Formula (Hill Notation):
C12H14O
CAS Number:
Molecular Weight:
174.24
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing