
Cycloolivil

Synonym(s):
(1S,2S,3S)-1,2,3,4-Tetrahydro-3,7-dihydroxy-1-(4-hydroxy-3-methoxyphenyl)-6-methoxy-2,3-naphthalenedimethanol, (2S,3S,4S)-4-(4-Hydroxy-3-methoxyphenyl)-2,3-bis(hydroxymethyl)-7-methoxy-1,2,3,4-tetrahydro-2,6-naphthalenediol, Isoolivil
Empirical Formula (Hill Notation):
C20H24O7
CAS Number:
Molecular Weight:
376.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing