
BENDAMUSTINE RCC

Synonym(s):
Ethyl 4-(5-(bis(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate
Empirical Formula (Hill Notation):
C18H27N3O4
CAS Number:
Molecular Weight:
349.42
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing